Assay Details
Gene Symbol: Slc4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009208.3 | NP_033234.2 | 11 - 12 | 1776 | 54 | |
GenBank mRNA | AK147218.1 | - | 2113 | 54 | ||
BC066086.1 | - | 12 - 13 | 1650 | 54 | ||
BC080271.1 | - | 11 - 12 | 1771 | 6831942 | 54 | |
M28383.1 | - | 11 - 12 | 1746 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
