Assay Details
Gene Symbol: Slc4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009208.3 | NP_033234.2 | 3 - 4 | 433 | 78 | |
GenBank mRNA | AK045583.1 | - | 2 - 3 | 320 | 78 | |
AK147218.1 | - | 445 | 78 | |||
BC080271.1 | - | 3 - 4 | 428 | 6831942 | 78 | |
M28383.1 | - | 3 - 4 | 403 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
