Assay Details
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148937.2 | NP_683739.2 | 10 - 11 | 1804 | 68 | |
GenBank mRNA | AK029561.1 | - | 10 - 11 | 1799 | 68 | |
AY033991.1 | - | 10 - 11 | 1636 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
