Assay Details
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281980.1 | NP_001268909.1 | 11 - 12 | 897 | 90 | |
NM_001281981.1 | NP_001268910.1 | 12 - 13 | 938 | 90 | ||
NM_008511.3 | NP_032537.3 | 11 - 12 | 1169 | 90 | ||
GenBank mRNA | AK153823.1 | - | 11 - 12 | 1167 | 90 | |
AK153865.1 | - | 11 - 12 | 899 | 90 | ||
AK160009.1 | - | 11 - 12 | 1129 | 90 | ||
AK166635.1 | - | 9 - 10 | 618 | 90 | ||
AK168775.1 | - | 9 - 10 | 595 | 90 | ||
AK169578.1 | - | 12 - 13 | 922 | 90 | ||
BC052909.1 | - | 11 - 12 | 1105 | 30076553 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
