Assay Details
Gene Symbol: Tuba3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009449.3 | NP_033475.1 | 1 - 2 | 124 | 58 | |
GenBank mRNA | BC050769.1 | - | 1 - 2 | 48 | 6771187 | 58 |
CK137160.1 | - | 1 - 2 | 124 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC050769.1, NM_009449.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
