Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400025401.mm.1 |
TC0400003206.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025401.mm.1 |
NM_001172073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025401.mm.1 |
NM_001172073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025401.mm.1 |
NM_028209.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025401.mm.1 |
NM_028209.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025403.mm.1 |
TC0400003206.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025403.mm.1 |
NM_001172073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025403.mm.1 |
NM_001172073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025403.mm.1 |
NM_028209.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025403.mm.1 |
NM_028209.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025406.mm.1 |
TC0400003206.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025406.mm.1 |
NM_001172073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025406.mm.1 |
NM_001172073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025406.mm.1 |
NM_028209.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025406.mm.1 |
NM_028209.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025405.mm.1 |
TC0400003206.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025405.mm.1 |
NM_001172073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025405.mm.1 |
NM_001172073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025405.mm.1 |
NM_028209.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025405.mm.1 |
NM_028209.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025404.mm.1 |
TC0400003206.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025404.mm.1 |
NM_001172073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025404.mm.1 |
NM_001172073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025404.mm.1 |
NM_028209.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025404.mm.1 |
NM_028209.2 |
Refseq |