Assay Details
Gene Symbol: Slc2a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085529.1 | NP_001078998.1 | 11 - 12 | 1357 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003603 | NM_001085529.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136781 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494457 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699069 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889601 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164384 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491822 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452637 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210320 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2526 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781128 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22446 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003603 | NM_001085529.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404561 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03574210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03986768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03813647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03612550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03746706 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03219079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03976017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04024021 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03219080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404589 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404587 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03619058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404583 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404584 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03219084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04035297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23404568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03986782 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10510544 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510534 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510535 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510538 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510542 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510533 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510537 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510543 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510536 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510541 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510545 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10510532 | - | - |
Affymetrix Mouse Clariom S Array | PSR0400016143.mm.1 | TC0400001971.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400016143.mm.1 | NM_001085529.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400016143.mm.1 | NM_001085529.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 349412 | - | - |
