Assay Details
Gene Symbol: Ccdc186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK041398.1 | - | 1 - 2 | 185 | 149 | |
AK054204.1 | - | 1 - 2 | 199 | 149 | ||
BC064039.1 | - | 1 - 2 | 161 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK041398.1, BC064039.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
