Assay Details
Gene Symbol: Ccdc186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170757.1 | NP_739563.1 | 1 - 2 | 686 | 150 | |
GenBank mRNA | AF349751.1 | - | 1 - 2 | 686 | 150 | |
AK041398.1 | - | 2 - 3 | 873 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
