Assay Details
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027976.2 | NP_082252.1 | 13 - 14 | 2043 | 67 | |
GenBank mRNA | AK006541.1 | - | 2 - 3 | 520 | 67 | |
AK034942.1 | - | 2043 | 67 | |||
AK145894.1 | - | 13 - 14 | 1704 | 67 | ||
AK150542.1 | - | 13 - 14 | 1709 | 67 | ||
AK150663.1 | - | 13 - 14 | 1709 | 67 | ||
AK151378.1 | - | 13 - 14 | 1498 | 67 | ||
AK161024.1 | - | 13 - 14 | 1396 | 67 | ||
AK168019.1 | - | 1 - 2 | 44 | 67 | ||
AK218456.1 | - | 2 - 3 | 216 | 67 | ||
BC031544.1 | - | 13 - 14 | 1477 | 3987201 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK006541.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
