Assay Details
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008149.3 | NP_032175.2 | 11 - 12 | 1373 | 67 | |
GenBank mRNA | AK047295.1 | - | 11 - 12 | 1374 | 67 | |
AK137067.1 | - | 13 - 14 | 1654 | 67 | ||
AK173202.1 | - | 11 - 12 | 1398 | 67 | ||
BC019201.1 | - | 11 - 12 | 1378 | 5041120 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
