Assay Details
Gene Symbol: Ensa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019561.2 | NP_062507.1 | 3 - 4 | 488 | 54 | |
GenBank mRNA | AK006149.1 | - | 3 - 4 | 458 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
