Assay Details
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122899.1 | NP_001116371.1 | 11 - 12 | 2294 | 106 | |
NM_010704.2 | NP_034834.1 | 11 - 12 | 2294 | 106 | ||
NM_146146.2 | NP_666258.2 | 11 - 12 | 2294 | 106 | ||
GenBank mRNA | AK140785.1 | - | 11 - 12 | 1796 | 106 | |
AK143989.1 | - | 11 - 12 | 1983 | 106 | ||
AK155891.1 | - | 11 - 12 | 1834 | 106 | ||
BC082551.1 | - | 11 - 12 | 1779 | 30621706 | 106 | |
U42467.1 | - | 11 - 12 | 1802 | 106 | ||
U46135.1 | - | 11 - 12 | 1802 | 106 | ||
U49106.1 | - | 10 - 11 | 1742 | 106 | ||
U49109.1 | - | 10 - 11 | 1742 | 106 | ||
U49110.1 | - | 10 - 11 | 1742 | 106 | ||
U52915.1 | - | 11 - 12 | 2256 | 106 | ||
U58861.1 | - | 11 - 12 | 1824 | 106 | ||
U58862.1 | - | 11 - 12 | 1834 | 106 | ||
U58863.1 | - | 11 - 12 | 1824 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
