Assay Details
Gene Symbol: Ndufs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160040.1 | NP_001153512.1 | 1 - 2 | 399 | 111 | |
GenBank mRNA | AK034597.1 | - | 1 - 2 | 310 | 111 | |
AK146814.1 | - | 1 - 2 | 259 | 111 | ||
BG808119.1 | - | 1 - 2 | 239 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK034597.1, AK146814.1, NM_001160040.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
