Assay Details
Gene Symbol: Pdgfrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146268.1 | NP_001139740.1 | 8 - 9 | 1666 | 79 | |
NM_008809.2 | NP_032835.2 | 8 - 9 | 1666 | 79 | ||
GenBank mRNA | AK134785.1 | - | 9 - 10 | 1688 | 79 | |
AK143247.1 | - | 8 - 9 | 1646 | 79 | ||
AK158145.1 | - | 2 - 3 | 323 | 79 | ||
AK203442.1 | - | 1 - 2 | 88 | 79 | ||
BC055311.1 | - | 8 - 9 | 1678 | 30060666 | 79 | |
DQ979390.1 | - | 2 - 3 | 322 | 79 | ||
X04367.1 | - | 8 - 9 | 1375 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
