Assay Details
Gene Symbol: Hsd17b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008292.4 | NP_032318.2 | 1 - 2 | 210 | 82 | |
GenBank mRNA | AK004866.1 | - | 1 - 2 | 210 | 82 | |
AK088381.1 | - | 1 - 2 | 185 | 82 | ||
AK147182.1 | - | 1 - 2 | 182 | 82 | ||
AK152532.1 | - | 1 - 2 | 105 | 82 | ||
AK161655.1 | - | 1 - 2 | 105 | 82 | ||
AK166801.1 | - | 1 - 2 | 123 | 82 | ||
AK167060.1 | - | 1 - 2 | 160 | 82 | ||
AK169077.1 | - | 1 - 2 | 112 | 82 | ||
BC022175.1 | - | 1 - 2 | 83 | 4209394 | 82 | |
X89998.1 | - | 1 - 2 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
