Assay Details
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172095.1 | NP_001165566.1 | 9 - 10 | 1280 | 153 | |
NM_144787.2 | NP_659036.1 | 9 - 10 | 1344 | 153 | ||
GenBank mRNA | AK042792.1 | - | 8 - 9 | 1059 | 153 | |
AK053104.1 | - | 9 - 10 | 1220 | 153 | ||
AK081769.1 | - | 9 - 10 | 1287 | 153 | ||
AK144103.1 | - | 9 - 10 | 1307 | 153 | ||
AK144890.1 | - | 9 - 10 | 1318 | 153 | ||
AK173023.1 | - | 9 - 10 | 1344 | 153 | ||
BC020180.1 | - | 9 - 10 | 1286 | 4923943 | 153 | |
BC042424.1 | - | 9 - 10 | 1280 | 5357211 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
