Assay Details
Gene Symbol: Dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027490.1 | NP_081766.1 | 10 - 11 | 1243 | 67 | |
GenBank mRNA | AK017809.1 | - | 10 - 11 | 1235 | 67 | |
AK152160.1 | - | 10 - 11 | 1216 | 67 | ||
AK153105.1 | - | 10 - 11 | 1245 | 67 | ||
AK153264.1 | - | 10 - 11 | 1216 | 67 | ||
AK153310.1 | - | 10 - 11 | 1216 | 67 | ||
AK166560.1 | - | 10 - 11 | 1222 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122564 | AK153264.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122564 | AK153310.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122564 | AK152160.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122564 | NM_027490.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62735 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552740 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428798 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227150 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469007 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414497 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277507 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231812 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752702 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67005 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325357 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737142 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805252 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447944 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683934 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224702 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120654 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153795 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973184 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46378 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100070 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12850 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74127 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161666 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92041 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154394 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175186 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122564 | AK153264.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122564 | AK153310.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122564 | AK152160.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122564 | NM_027490.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154394 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08984100 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08984192 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08984118 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08984179 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175186 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03463699 | - | - |
Whole Mouse Genome G4122A | A_51_P190791 | AK166560.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P190791 | AK153105.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P190791 | NM_027490.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P190791 | AK008609.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P190791 | AK010490.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P678833 | AK166560.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P678833 | AK153264.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P678833 | AK153310.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P678833 | AK152160.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P678833 | NM_027490.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175186 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154394 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08984118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666582 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13698786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666538 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13856995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666506 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13813845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13986776 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13767383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666554 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13862875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666399 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13955330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666515 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666542 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13900407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13926948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13956977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13767385 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14139897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13862867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14039682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14114482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666390 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666447 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666546 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666556 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666429 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14106997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14044436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14070235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666517 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666495 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13845359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14106988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13986770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14106991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666425 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666540 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666587 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666553 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13978725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666512 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13862865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666584 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666403 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666402 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14106986 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13862861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31666423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13978724 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10455475 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455482 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455478 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455480 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455479 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455477 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455474 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455481 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430150_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455070_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10455472 | - | - |
Mouse Genome 430 2.0 Array | 1455070_at | - | - |
Mouse Genome 430 2.0 Array | 1430150_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430150_at | - | - |
Mouse Genome 430 2.0 Array | 1455070_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70640_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70640_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800003366.mm.1 | NM_027490.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003366.mm.1 | NM_027490.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003366.mm.1 | TC1800000457.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800003365.mm.1 | NM_027490.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003365.mm.1 | NM_027490.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003365.mm.1 | TC1800000457.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800003368.mm.1 | NM_027490.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003368.mm.1 | NM_027490.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003368.mm.1 | TC1800000457.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800003367.mm.1 | NM_027490.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003367.mm.1 | NM_027490.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003367.mm.1 | TC1800000457.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455070_at | - | - |
Mouse Expression Set 430 | 1430150_PM_at | - | - |
Mouse Expression Set 430 | 1430150_at | - | - |
Mouse Expression Set 430 | 1455070_PM_at | - | - |
Murine Genome U74v2 Set | 135023_PM_at | - | - |
Murine Genome U74v2 Set | 135023_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430150_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455070_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455070_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430150_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 635328 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2704951 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2704951 | - | - |
