Assay Details
Gene Symbol: Oacyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177028.3 | NP_796002.1 | 7 - 8 | 1037 | 95 | |
GenBank mRNA | AK030604.1 | - | 7 - 8 | 1037 | 95 | |
AK030615.1 | - | 5 - 6 | 817 | 95 | ||
BC141265.1 | - | 7 - 8 | 1034 | 9055877 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
