Assay Details
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080555.2 | NP_542122.1 | 4 - 5 | 1116 | 65 | |
GenBank mRNA | AK011276.1 | - | 4 - 5 | 1115 | 65 | |
AK076183.1 | - | 4 - 5 | 812 | 65 | ||
AK159136.1 | - | 4 - 5 | 1202 | 65 | ||
AK160056.1 | - | 4 - 5 | 1016 | 65 | ||
BC005558.1 | - | 4 - 5 | 1103 | 3964817 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
