Assay Details
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252458.1 | NP_001239387.1 | 3 - 4 | 653 | 90 | |
NM_178900.4 | NP_849231.1 | 2 - 3 | 938 | 90 | ||
GenBank mRNA | AK037030.1 | - | 2 - 3 | 939 | 90 | |
AK041207.1 | - | 1 - 2 | 158 | 90 | ||
AK154029.1 | - | 2 - 3 | 787 | 90 | ||
AK170236.1 | - | 3 - 4 | 626 | 90 | ||
AK170761.1 | - | 3 - 4 | 617 | 90 | ||
BC095949.1 | - | 3 - 4 | 603 | 4950957 | 90 | |
BC096444.1 | - | 3 - 4 | 621 | 4216827 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
