Assay Details
Gene Symbol: Gm15704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029019.1 | - | 91 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
tubulin, alpha 3B |
Tuba3b |
M[a]7, Tuba7 |
-124.0kb | Anti-sense | Chr. 6 - 145615963 - 145621477 on NCBI Build GRCm38 |
Downstream | Ras association (RalGDS/AF-6) domain family (N-terminal) member 8 | Rassf8 |
5133400D11Rik, AA815821, AW123240, Hoj1, mHoj-1 |
292.0 | Anti-sense | Chr. 6 - 145746562 - 145821079 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
