Assay Details
Gene Symbol: 1700047F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006706.1 | - | 1 - 2 | 347 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
DMRT-like family B with proline-rich C-terminal, 1 |
Dmrtb1 |
Dmrt6, Prp13 |
-22.8kb | Anti-sense | Chr. 4 - 107676288 - 107684243 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4933424M12 gene | 4933424M12Rik | 68.4kb | Sense | Chr. 4 - 107783708 - 107789922 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
