Assay Details
Gene Symbol: 1700047F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006706.1 | - | 2 - 3 | 466 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
DMRT-like family B with proline-rich C-terminal, 1 |
Dmrtb1 |
Dmrt6, Prp13 |
-31.0kb | Anti-sense | Chr. 4 - 107676288 - 107684243 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4933424M12 gene | 4933424M12Rik | 66.8kb | Sense | Chr. 4 - 107783708 - 107789922 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020716 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207299 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109234 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109234 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140850 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03647940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03390653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03640507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03780271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03390662 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03445783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03626825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03405087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03993377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192374 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03509683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192383 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03982593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03835339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03701179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03876489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03458701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03509680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03647939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03701177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03390648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03598463 | - | - |
