Assay Details
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178143.2 | NP_835279.2 | 3 - 4 | 425 | 96 | |
GenBank mRNA | AK044030.1 | - | 3 - 4 | 261 | 96 | |
AK134573.1 | - | 3 - 4 | 283 | 96 | ||
BC138565.1 | - | 3 - 4 | 434 | 8861585 | 96 | |
BC138566.1 | - | 3 - 4 | 434 | 8861586 | 96 | |
BY718361.1 | - | 3 - 4 | 280 | 96 | ||
FJ588504.1 | - | 2 - 3 | 183 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
