Assay Details
Gene Symbol: Plekhg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285999.1 | NP_001272928.1 | 18 - 19 | 2325 | 78 | |
NR_104381.1 | 19 - 20 | 2291 | 78 | |||
GenBank mRNA | AK129198.1 | - | 5 - 6 | 533 | 78 | |
AK138147.1 | - | 4 - 5 | 1273 | 78 | ||
AK154912.1 | - | 19 - 20 | 2284 | 78 | ||
AY605057.1 | - | 18 - 19 | 2325 | 78 | ||
AY605058.1 | - | 18 - 19 | 2325 | 78 | ||
BC023181.1 | - | 4 - 5 | 471 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
