Assay Details
Gene Symbol: Plekhg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285999.1 | NP_001272928.1 | 19 - 20 | 2507 | 90 | |
NR_104381.1 | 20 - 21 | 2473 | 90 | |||
GenBank mRNA | AK129198.1 | - | 6 - 7 | 715 | 90 | |
AK138147.1 | - | 5 - 6 | 1455 | 90 | ||
AK154912.1 | - | 20 - 21 | 2466 | 90 | ||
AY605057.1 | - | 19 - 20 | 2507 | 90 | ||
AY605058.1 | - | 19 - 20 | 2507 | 90 | ||
BC023181.1 | - | 5 - 6 | 653 | 90 | ||
BC064091.1 | - | 1 - 2 | 108 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
