Assay Details
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008720.2 | NP_032746.2 | 21 - 22 | 3393 | 100 | |
GenBank mRNA | AF003348.1 | - | 21 - 22 | 3370 | 100 | |
BC052437.1 | - | 21 - 22 | 3393 | 6405214 | 100 | |
BC054539.1 | - | 21 - 22 | 3393 | 6405214 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
