Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
NM_001005787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
NM_001005787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
NM_007704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
NM_007704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006623.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
NM_001005787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
NM_001005787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
NM_007704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
NM_007704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006622.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006617.mm.1 |
NM_001005787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006617.mm.1 |
NM_001005787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006617.mm.1 |
NM_007704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006617.mm.1 |
NM_007704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006617.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006619.mm.1 |
NM_001005787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006619.mm.1 |
NM_001005787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006619.mm.1 |
NM_007704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006619.mm.1 |
NM_007704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006619.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006618.mm.1 |
NM_001005787.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006618.mm.1 |
NM_001005787.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006618.mm.1 |
NM_007704.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006618.mm.1 |
NM_007704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006618.mm.1 |
TC0400000982.mm.1 |
MTA TC |