Assay Details
Gene Symbol: Cog5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163126.1 | NP_001156598.1 | 17 - 18 | 1877 | 79 | |
GenBank mRNA | AK030656.1 | - | 17 - 18 | 1878 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
