Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400017248.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017248.mm.1 |
NM_001159637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017248.mm.1 |
NM_001159637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017253.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017253.mm.1 |
NM_001159637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017253.mm.1 |
NM_001159637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017253.mm.1 |
NM_138671.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017253.mm.1 |
NM_138671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017255.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017255.mm.1 |
NM_001159637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017255.mm.1 |
NM_001159637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017255.mm.1 |
NM_138671.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017255.mm.1 |
NM_138671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017243.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017243.mm.1 |
NM_138671.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017243.mm.1 |
NM_138671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017256.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017256.mm.1 |
NM_001159637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017256.mm.1 |
NM_001159637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017256.mm.1 |
NM_138671.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017256.mm.1 |
NM_138671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017257.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017257.mm.1 |
NM_001159637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017257.mm.1 |
NM_001159637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017257.mm.1 |
NM_138671.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017257.mm.1 |
NM_138671.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017247.mm.1 |
TC0400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017247.mm.1 |
NM_001159637.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017247.mm.1 |
NM_001159637.1 |
Refseq |