Assay Details
Gene Symbol: Nadk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159637.1 | NP_001153109.1 | 4 - 5 | 583 | 79 | |
NM_138671.2 | NP_619612.2 | 4 - 5 | 482 | 79 | ||
GenBank mRNA | AK147917.1 | - | 4 - 5 | 560 | 79 | |
AK164862.1 | - | 2 - 3 | 667 | 79 | ||
AK165510.1 | - | 4 - 5 | 576 | 79 | ||
AK170095.1 | - | 4 - 5 | 564 | 79 | ||
AK170608.1 | - | 4 - 5 | 578 | 79 | ||
AK170960.1 | - | 4 - 5 | 564 | 79 | ||
BC004012.1 | - | 4 - 5 | 470 | 3493623 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
