Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10511011 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10511008 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10511010 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10511012 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1456310_PM_a_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1438154_PM_x_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1431003_PM_a_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10511007 |
- |
- |
Mouse Genome 430 2.0 Array |
1438154_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431003_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431003_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456310_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456310_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1438154_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1442780_at |
- |
- |
Mouse Genome 430 2.0 Array |
1442780_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
67513_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
67513_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0400017096.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017096.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017096.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017098.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017098.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017098.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017104.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017104.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017104.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017104.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017104.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017114.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017114.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017114.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017114.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017114.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017116.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017116.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017116.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017116.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017116.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017107.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017107.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017107.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017107.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017107.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017112.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017112.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017112.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017112.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017112.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017101.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017101.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017101.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017101.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017101.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017092.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017092.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017092.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017092.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017092.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017109.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017109.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017109.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017109.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017109.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017110.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017110.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017110.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017110.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017110.mm.1 |
NR_033802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017108.mm.1 |
TC0400002069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017108.mm.1 |
NM_001190445.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017108.mm.1 |
NM_001190445.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017108.mm.1 |
NR_033802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017108.mm.1 |
NR_033802.1 |
Refseq |
Mouse Expression Set 430 |
1438154_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1431003_a_at |
- |
- |
Mouse Expression Set 430 |
1456310_a_at |
- |
- |
Mouse Expression Set 430 |
1456310_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1431003_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1438154_x_at |
- |
- |
Murine Genome U74v2 Set |
102056_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
102057_r_at |
- |
- |
Murine Genome U74v2 Set |
102056_f_at |
- |
- |
Murine Genome U74v2 Set |
102057_PM_r_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1431003_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1431003_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1438154_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456310_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456310_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1438154_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1438154_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1438154_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456310_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1431003_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1431003_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456310_a_at |
- |
- |