Assay Details
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256107.1 | NP_001243036.1 | 11 - 12 | 2221 | 68 | |
NM_001256108.2 | NP_001243037.2 | 11 - 12 | 2041 | 68 | ||
NM_145124.3 | NP_660106.2 | 10 - 11 | 2125 | 68 | ||
GenBank mRNA | AK084791.1 | - | 3 - 4 | 423 | 68 | |
AK147940.1 | - | 11 - 12 | 1986 | 68 | ||
AK159611.1 | - | 11 - 12 | 1938 | 68 | ||
BC058086.1 | - | 9 - 10 | 1613 | 68 | ||
BC141133.1 | - | 8 - 9 | 1514 | 9055745 | 68 | |
BC145441.1 | - | 9 - 10 | 1610 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
