Assay Details
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037859.2 | NP_001032948.2 | 14 - 15 | 2246 | 116 | |
GenBank mRNA | AK004947.1 | - | 13 - 14 | 2056 | 116 | |
AK041299.1 | - | 13 - 14 | 2034 | 116 | ||
AK076215.1 | - | 13 - 14 | 2055 | 116 | ||
AK079247.1 | - | 13 - 14 | 2063 | 116 | ||
AK080326.1 | - | 13 - 14 | 2044 | 116 | ||
AK143545.1 | - | 13 - 14 | 2047 | 116 | ||
AK154128.1 | - | 13 - 14 | 2061 | 116 | ||
AK154556.1 | - | 13 - 14 | 2057 | 116 | ||
AK154653.1 | - | 13 - 14 | 2057 | 116 | ||
AK154699.1 | - | 13 - 14 | 2057 | 116 | ||
AK155488.1 | - | 13 - 14 | 2067 | 116 | ||
AK155576.1 | - | 2063 | 116 | |||
AK155640.1 | - | 13 - 14 | 2061 | 116 | ||
AK155654.1 | - | 13 - 14 | 2061 | 116 | ||
AK155858.1 | - | 13 - 14 | 2064 | 116 | ||
BC036343.1 | - | 13 - 14 | 2033 | 4948135 | 116 | |
BC043054.1 | - | 13 - 14 | 2037 | 6418718 | 116 | |
BC066863.1 | - | 14 - 15 | 2155 | 30436119 | 116 | |
X68932.1 | - | 13 - 14 | 1949 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
