Assay Details
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037859.2 | NP_001032948.2 | 6 - 7 | 1172 | 71 | |
GenBank mRNA | AK004947.1 | - | 5 - 6 | 982 | 71 | |
AK041299.1 | - | 5 - 6 | 960 | 71 | ||
AK076215.1 | - | 5 - 6 | 980 | 71 | ||
AK079247.1 | - | 5 - 6 | 989 | 71 | ||
AK080326.1 | - | 5 - 6 | 969 | 71 | ||
AK089498.1 | - | 5 - 6 | 962 | 71 | ||
AK143545.1 | - | 5 - 6 | 973 | 71 | ||
AK146069.1 | - | 6 - 7 | 1173 | 71 | ||
AK154128.1 | - | 5 - 6 | 987 | 71 | ||
AK154556.1 | - | 5 - 6 | 983 | 71 | ||
AK154653.1 | - | 5 - 6 | 983 | 71 | ||
AK154699.1 | - | 5 - 6 | 983 | 71 | ||
AK155488.1 | - | 5 - 6 | 992 | 71 | ||
AK155565.1 | - | 5 - 6 | 1002 | 71 | ||
AK155576.1 | - | 989 | 71 | |||
AK155640.1 | - | 5 - 6 | 987 | 71 | ||
AK155654.1 | - | 5 - 6 | 987 | 71 | ||
AK155858.1 | - | 5 - 6 | 990 | 71 | ||
AK171241.1 | - | 5 - 6 | 1022 | 71 | ||
AK171386.1 | - | 5 - 6 | 987 | 71 | ||
AK171543.1 | - | 5 - 6 | 962 | 71 | ||
BC036343.1 | - | 5 - 6 | 959 | 4948135 | 71 | |
BC043054.1 | - | 5 - 6 | 963 | 6418718 | 71 | |
BC050024.1 | - | 5 - 6 | 1029 | 71 | ||
BC066863.1 | - | 6 - 7 | 1081 | 30436119 | 71 | |
X06368.1 | - | 5 - 6 | 965 | 71 | ||
X68932.1 | - | 5 - 6 | 875 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
