Assay Details
Gene Symbol: Mroh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126487.1 | NP_001119959.1 | 20 - 21 | 3591 | 123 | |
GenBank mRNA | AK134749.1 | - | 11 - 12 | 2013 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
