Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400025465.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025465.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025465.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025453.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025453.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025453.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025459.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025459.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025459.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025462.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025462.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025462.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025455.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025455.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025455.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025461.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025461.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025461.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025457.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025457.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025457.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025463.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025463.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025463.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025464.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025464.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025464.mm.1 |
NM_001126487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025466.mm.1 |
TC0400003207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025466.mm.1 |
NM_001126487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025466.mm.1 |
NM_001126487.1 |
Refseq |