Assay Details
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC005464.1 | - | 1 - 2 | 37 | 3591420 | 72 |
BC011160.1 | - | 1 - 2 | 64 | 4024353 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC005464.1, BC011160.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
