Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
TC0200002150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_001252543.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_001252543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_001252544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_001252544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_001252545.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_001252545.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_019787.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016980.mm.1 |
NM_019787.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
TC0200002150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_001252543.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_001252543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_001252544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_001252544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_001252545.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_001252545.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_019787.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016964.mm.1 |
NM_019787.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
TC0200002150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_001252543.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_001252543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_001252544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_001252544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_001252545.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_001252545.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_019787.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016978.mm.1 |
NM_019787.4 |
Refseq |