Assay Details
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172430.3 | NP_766018.3 | 4 - 5 | 488 | 70 | |
GenBank mRNA | AK020841.1 | - | 5 - 6 | 463 | 70 | |
AK038638.1 | - | 5 - 6 | 533 | 70 | ||
BC069832.1 | - | 4 - 5 | 454 | 30431760 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
