Assay Details
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290649.1 | NP_001277578.1 | 15 - 16 | 1760 | 88 | |
NM_029793.2 | NP_084069.1 | 16 - 17 | 1835 | 88 | ||
NR_110968.1 | 16 - 17 | 1835 | 88 | |||
GenBank mRNA | AK028516.1 | - | 16 - 17 | 1755 | 88 | |
AK044987.1 | - | 15 - 16 | 2245 | 88 | ||
BC037037.1 | - | 11 - 12 | 1171 | 88 | ||
BC043090.1 | - | 16 - 17 | 1700 | 6418353 | 88 | |
BC053068.1 | - | 15 - 16 | 1688 | 6405595 | 88 | |
BC080733.1 | - | 15 - 16 | 1760 | 30110235 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
