Assay Details
Gene Symbol: Arrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177231.2 | NP_796205.1 | 5 - 6 | 643 | 67 | |
NM_178220.3 | NP_835738.1 | 5 - 6 | 643 | 67 | ||
GenBank mRNA | AK052588.1 | - | 5 - 6 | 420 | 67 | |
AK090090.1 | - | 5 - 6 | 470 | 67 | ||
AK144054.1 | - | 5 - 6 | 442 | 67 | ||
AK147508.1 | - | 5 - 6 | 644 | 67 | ||
AK147566.1 | - | 5 - 6 | 665 | 67 | ||
AK165514.1 | - | 5 - 6 | 442 | 67 | ||
BC108969.1 | - | 5 - 6 | 410 | 40047224 | 67 | |
BC108970.1 | - | 5 - 6 | 410 | 40047225 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
