Assay Details
Gene Symbol: Tacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004468.2 | NP_001004468.1 | 19 - 20 | 8347 | 64 | |
NM_021314.4 | NP_067289.2 | 15 - 16 | 3182 | 64 | ||
NM_206856.3 | NP_996738.2 | 18 - 19 | 3148 | 64 | ||
GenBank mRNA | AB041546.1 | - | 15 - 16 | 3180 | 64 | |
AK145721.1 | - | 13 - 14 | 2811 | 64 | ||
AK165194.1 | - | 14 - 15 | 3006 | 64 | ||
AK165603.1 | - | 12 - 13 | 2706 | 64 | ||
AK220527.1 | - | 1 - 2 | 2985 | 64 | ||
AY177410.1 | - | 17 - 18 | 3157 | 64 | ||
BC004057.1 | - | 10 - 11 | 2222 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
