Assay Details
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172671.2 | NP_766259.2 | 2 - 3 | 732 | 95 | |
GenBank mRNA | AY255619.1 | - | 1 - 2 | 72 | 95 | |
BE987673.1 | - | 2 - 3 | 269 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
