Assay Details
Gene Symbol: Ddx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134040.1 | NP_598801.1 | 19 - 20 | 1644 | 150 | |
GenBank mRNA | AK028341.1 | - | 19 - 20 | 1641 | 150 | |
AK132157.1 | - | 19 - 20 | 1684 | 150 | ||
AK153335.1 | - | 19 - 20 | 1668 | 150 | ||
AK160982.1 | - | 19 - 20 | 1645 | 150 | ||
AK166146.1 | - | 19 - 20 | 1627 | 150 | ||
BC008570.1 | - | 19 - 20 | 1622 | 3587491 | 150 | |
BC010624.1 | - | 19 - 20 | 1644 | 3153029 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
