Assay Details
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145529.3 | NP_663504.1 | 16 - 17 | 1639 | 132 | |
GenBank mRNA | AK159180.1 | - | 16 - 17 | 1442 | 132 | |
AK160306.1 | - | 2 - 3 | 175 | 132 | ||
BC003241.1 | - | 16 - 17 | 1600 | 3498424 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
