Assay Details
Gene Symbol: Nol10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008421.1 | NP_001008421.1 | 4 - 5 | 354 | 151 | |
GenBank mRNA | AK148210.1 | - | 4 - 5 | 370 | 151 | |
AK160864.1 | - | 4 - 5 | 390 | 151 | ||
AK164400.1 | - | 4 - 5 | 380 | 151 | ||
AK166160.1 | - | 4 - 5 | 383 | 151 | ||
BC086676.1 | - | 4 - 5 | 354 | 30659737 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
