Assay Details
Gene Symbol: Ppp1r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163818.1 | NP_001157290.1 | 13 - 14 | 2039 | 71 | |
GenBank mRNA | AJ504718.1 | - | 13 - 14 | 2045 | 71 | |
AK145485.1 | - | 12 - 13 | 1483 | 71 | ||
BC029765.1 | - | 5 - 6 | 713 | 71 | ||
BC052059.1 | - | 13 - 14 | 2042 | 5702171 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
