Assay Details
Gene Symbol: Pde3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011055.2 | NP_035185.2 | 13 - 14 | 2948 | 147 | |
GenBank mRNA | AF547435.1 | - | 13 - 14 | 2921 | 147 | |
AJ132271.1 | - | 13 - 14 | 2651 | 147 | ||
X95521.1 | - | 13 - 14 | 1793 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
